missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myb Antibody (MYB286), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34690B
This item is not returnable.
View return policy
Description
c-Myb Monoclonal specifically detects c-Myb in Human samples. It is validated for ELISA.
Specifications
| c-Myb | |
| Monoclonal | |
| Biotin | |
| c-myb, Cmyb, c-myb protein (140 AA), c-myb_CDS, c-myb10A_CDS, c-myb13A_CDS, c-myb14A_CDS, c-myb8B_CDS, efg, Proto-oncogene c-Myb, transcriptional activator Myb, v-myb avian myeloblastosis viral oncogene homolog, v-myb myeloblastosis viral oncogene homolog (avian) | |
| Mouse | |
| 75 kDa | |
| 0.1 mL | |
| Cancer | |
| 4602 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| MYB286 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| MYB | |
| A synthetic peptide, corresponding to aa 119-135 (RRKVEQEGYPQESSKAG) of c-myb onco protein, coupled to KLH (Uniprot: P01104) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The highly leukemogenic avian retrovirus E26 contains two oncogenes, v-Myb and v-Ets, which are expressed together as a fusion protein. The cellular homolog of v-Myb, designated c-Myb, encodes a transcription factor. Deletion or disruption of a negative regulatory domain mapping within the carboxy-terminal domain of c-Myb results in enhanced transactivating capacity and in parallel, leads to activation of its ability to transform hemopoietic cells. c-Myb is expressed preferentially, but not exclusively, in immature hemopoietic cells and its expression decreases as cells differentiate. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction