missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Mpl Recombinant Rabbit Monoclonal Antibody (K01_2J43)

Product Code. 30830311
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830311 50 μL
30825621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830311 Supplier Invitrogen™ Supplier No. MA565341
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

c-Mpl Recombinant Monoclonal Antibody for Western Blot

In 1990 an oncogene, v-mpl, was identified from the murine myeloproliferative leukemia virus that was capable of immortalizing bone marrow hematopoietic cells from different lineages. In 1992 the human homologue, named, c-mpl, was cloned. Sequence data revealed that c-mpl encoded a protein that was homologous with members of the hematopoietic receptor superfamily. Presence of anti-sense oligodeoxynucleotides of c-mpl inhibited megakaryocyte colony formation. The ligand for c-mpl, thrombopoietin, was cloned in 1994. Thrombopoietin was shown to be the major regulator of megakaryocytopoiesis and platelet formation. The protein encoded by the c-mpl gene, CD110, is a 635 amino acid transmembrane domain, with two extracellular cytokine receptor domains and two intracellular cytokine receptor box motifs. TPO-R deficient mice were severely thrombocytopenic, emphasizing the important role of CD110 and thrombopoietin in megakaryocyte and platelet formation. Upon binding of thrombopoietin CD110 is dimerized and the JAK family of non-receptor tyrosine kinases, as well as the STAT family, the MAPK family, the adaptor protein Shc and the receptors themselves become tyrosine phosphorylated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Mpl
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2J43
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene MPL
Gene Accession No. P40238, Q08351
Gene Alias CD110; c-mpl; hlb219; MPL; MPL proto-oncogene, thrombopoietin receptor; MPLV; myeloproliferative leukemia protein; myeloproliferative leukemia virus oncogene; proto-oncogene c-Mpl; THCYT2; thrombopoietin receptor; TPOR; TPO-R
Gene Symbols MPL
Host Species Rabbit
Immunogen A synthetic peptide of human TPOR.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17480, 366455, 4352
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.