Learn More
Olympus™ C-Mount Camera Adapter
Product Code. 15872445
Change view
Click to view available options
For Use With (Equipment):
Microscope
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15872445 Microscope Each
1 options
Product Code. 15872445 Supplier Olympus™ Supplier No. N5687400
  • ONLINE EXCLUSIVE PRICE
    £1718.88
    £1209.90 / Pack of 1
    Save £508.98
    29% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Facilitates use of camera to record images observed under your microscope.
  • C-mount.
  • 0.5x magnification gives full field number (FN) 22 on 2/3 in. CCD camera chips.
  • High IR transmission.
Specifications
Type C-mount Camera
Description 0.5X magnification
For Use With (Equipment) Microscope