Learn More
Medline Scientific™ C-Mount Camera Adapter
These adapters connect the camera to the microscope, ensuring that par focality is retained.
Brand:  Medline Scientific™ 7357.1111
Product Code. 12078598
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Provides for the connection of a digital camera to the microscope
  • Places the camera chip in the same focal plan as the eyepieces so that no re-focusing is required when switching between manual or on-screen viewing