missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Met Recombinant Rabbit Monoclonal Antibody (6O7V1)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557867

 View more versions of this product

Product Code. 30284487

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QIKDLGSELV RYDARVHTPH LDRLVSARSV SPTTEMVSNE SVDYRATFPE DQFPNSSQNG SCRQVQYPLT DMSPILTSGD SDISSPLLQN TVHIDLSALN PELVQAVQHV VIGPSSLIVH FNEVIGRGHF GCVYHGTLLD NDGKKIHCAV KSLNRITDIG EVSQFLTEGI IMKDFSHPNV LSLLGICLRS EGSPLVVLPY MKHGDLRNFI RNETHNPTVK DLIGFGLQVA KGMKYLASKK FVHRDLAARN CMLDEKFTVK VADFGLARDM YDKEYYSVHN KTGAKLPVKW MALESLQTQK FT.

MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

c-Met
Recombinant Monoclonal
1 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P08581, P16056
MET
Recombinant protein (or fragment).
20 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Western Blot, Immunocytochemistry
6O7V1
Unconjugated
MET
AI838057; AUTS9; bHLHe59; CG1705; CG1705-PA; CG1705-PB; cmet; c-met; c-met proto-oncogene; c-Met receptor tyrosine kinase; c-Met/HGF receptor; D249; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; HGFR; HGF-SF receptor; juvenile hormone resistance; met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; Met; protein tyrosin kinase; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; Proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine-protein kinase Met
Rabbit
Protein A
RUO
17295, 4233
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.