Learn More
Invitrogen™ c-Met Recombinant Rabbit Monoclonal Antibody (6O7V1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557868
Description
Immunogen sequence: QIKDLGSELV RYDARVHTPH LDRLVSARSV SPTTEMVSNE SVDYRATFPE DQFPNSSQNG SCRQVQYPLT DMSPILTSGD SDISSPLLQN TVHIDLSALN PELVQAVQHV VIGPSSLIVH FNEVIGRGHF GCVYHGTLLD NDGKKIHCAV KSLNRITDIG EVSQFLTEGI IMKDFSHPNV LSLLGICLRS EGSPLVVLPY MKHGDLRNFI RNETHNPTVK DLIGFGLQVA KGMKYLASKK FVHRDLAARN CMLDEKFTVK VADFGLARDM YDKEYYSVHN KTGAKLPVKW MALESLQTQK FT.
MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.
Specifications
| c-Met | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P08581, P16056 | |
| MET | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 6O7V1 | |
| Unconjugated | |
| MET | |
| AI838057; AUTS9; bHLHe59; CG1705; CG1705-PA; CG1705-PB; cmet; c-met; c-met proto-oncogene; c-Met receptor tyrosine kinase; c-Met/HGF receptor; D249; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; HGFR; HGF-SF receptor; juvenile hormone resistance; met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; Met; protein tyrosin kinase; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; Proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine-protein kinase Met | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17295, 4233 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.