missing translation for 'onlineSavingsMsg'
Learn More

c-Met inhibitor 1, MedChemExpress

Product Code. p-200099484
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489327

Brand: Medchem Express HY15735

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress c-Met inhibitor 1 is an inhibitor of the c-Met receptor signaling pathway useful for the treatment of cancer including gastric, glioblastoma, and pancreatic cancer.

c-Met inhibitor 1 is an inhibitor of the c-Met receptor signaling pathway useful for the treatment of cancer including gastric, glioblastoma, and pancreatic cancer.

Specifications

Chemical Name or Material c-Met inhibitor 1
Physical Form Solid
CAS 1357072-61-7
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H14N8S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (55.19 mM; Need ultrasonic)
SMILES CN1N=CC(C2=NN3C(C=C2)=NN=C3SC4=CC5=CN(C)N=C5C=C4)=C1
Molecular Weight (g/mol) 362.41
Formula Weight 362.41
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.