missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Maf Antibody (JE39-57), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32197
This item is not returnable.
View return policy
Description
c-Maf Monoclonal antibody specifically detects c-Maf in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| c-Maf | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, c-MAF, c-maf proto-oncogene, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian) | |
| Synthetic peptide within Human c-Maf aa 324-373/ 373. (Uniprot: O75444) | |
| 100 μL | |
| Apoptosis, Cancer, Cell Biology, Immunology | |
| 4094 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE39-57 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction