missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ C-Laurdan
GREENER_CHOICE

Product Code. 18747146
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 18747146

Brand: Tocris Bioscience™ 7273/2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Two-photon polarity-sensitive lipid membrane probe

C-Laurdan is a fluorescent probe for imaging lipid rafts. Used to detect membrane polarity via one and two-photon fluorecence imaging. Two photon excitation spectra = 780 nm; one photon absorption/emission spectra = 348/423 nm; Quantum yield = 0.43.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methyl-N-[6-(1-oxododecyl)-2-naphthalenyl]glycine
CAS 959839-06-6
Quantity 2 mg
Target Fluorescent Lipids Probes and Cell Membrane Stains
Molecular Formula C25H35NO3
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.