Learn More
Eurofins DiscoverX c-Kit Protein, Active
Description
Binding of stem cell factor (SCF) to C-Kit receptor tyrosine kinase induces homodimerization and intermolecular tyrosine phosphorylation of the receptor, creating docking sites for a number of SH2-containing signal transduction molecules. These include phosphatidylinositol 3'-kinase (PI 3K) and phospholipase C-1, Stat1, the Src family kinases Src, Lyn, and Fyn, and the GTPase activating protein GAP. Mutation in Kit which result in ligand-independent activation of the kinase were found to be associated with human gastrointestinal stromal tumors and mastocytosis.
GenBank X06182
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 76.7 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥67% |
| Protein | Kit, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.