missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Kit (D816V Mutant) Recombinant Rabbit Monoclonal Antibody (HL2653)
GREENER_CHOICE

Product Code. 30276272
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30276272 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30276272 Supplier Invitrogen™ Supplier No. MA556652

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

c-Kit (D816V Mutant) Recombinant Monoclonal Antibody for Western Blot, ICC/IF

This gene encodes the human homolog of the proto-oncogene c-kit C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit This protein is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor) Mutations in this gene are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous lukemia, and piebaldism Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Kit (D816V Mutant)
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2653
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Accession No. P29466, P48729, Q8N752
Gene Alias CD117; C-Kit; KIT proto-oncogene, receptor tyrosine kinase; MASTC; PBT; SCFR
Host Species Rabbit
Immunogen Synthetic peptide of human c-Kit (D816V Mutant).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011, 1452, 834
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.