missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Jun (Total) Multispecies InstantOne™ ELISA Kit

Product Code. 15550807
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 15550807

Brand: Invitrogen™ 858616111

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • User Manual
  • InstantOne™ Assay Plate - 96 well (sealed)
  • Cell Lysis Buffer (5X)
  • Enhancer Solution
  • Wash Buffer (10X)
  • c-Jun (Total) Capture Antibody Reagent
  • c-Jun (Total) Detection Antibody Reagent
  • Positive Control Cell Lysate (lyophilized)
  • Detection Reagent (TMB Substrate)
  • Stop Solution
  • Plate Seal

InstantOne ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of total human and mouse c-Jun in cell lysates. Detection of rat c-Jun is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

Fos and Jun dimerize to form Activator Protein-1 (AP-1), a transcriptional factor that binds to the 12-O-tetradecanoylphorbol 13-acetate (TPA) response element (TRE) of several cellular and viral genes including human collagenase, metallothionein IIa, stromelysin, interleukin 2, SV40 and polyoma. Fos and Jun contain the ′leucine-zipper′ motif that allows for dimerization and an adjacent basic domain required for biological activity. The functionally active form of Fos is in a heterodimer with a member of the Jun family. While Jun family members can form functional homodimers, studies indicate that Fos family members do not self-associate and therefore do not bind DNA on their own. The various dimers differ in their ability to transactivate AP-1 dependent genes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05412,P05627
Conjugate Biotin
Product Type ELISA Kit
Sample Type Cell Lysates
Target Species Human, Mouse
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 16476, 3725
Gene Symbol AP-1,AP1,c-Jun
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate,20 μL
Storage Requirements 2°C to 8°C
Synonym JUN, Transcription factor AP-1
Target Human, Mouse
Test Time 1 hr. 20 min.
Total Assay Time 1 hr. 10 min.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.