missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-jun [p Thr91, p Thr93] Antibody (C-J 4C4/1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2748CL1
This item is not returnable.
View return policy
Description
c-jun Monoclonal antibody specifically detects c-jun in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| c-jun | |
| Monoclonal | |
| CoraFluor 1 | |
| Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
| C-Jun phosphorylated at T91 and T93 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
| 3725 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Western Blot | |
| C-J 4C4/1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction