missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-jun, p Ser63 Antibody (S07-3G7), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £501.00
Specifications
| Antigen | c-jun |
|---|---|
| Clone | S07-3G7 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry 1:20, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654075
|
Novus Biologicals
NBP3-20024-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641555
|
Novus Biologicals
NBP3-20024-100ul |
100 μg |
£501.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-jun Monoclonal antibody specifically detects c-jun in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| c-jun | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 3725 | |
| IgG | |
| Affinity purified |
| S07-3G7 | |
| Western Blot, ELISA, Immunohistochemistry 1:20, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
| A synthetic phosphopeptide corresponding to residues surrounding Ser63 of human c-jun (Uniprot # P05412) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title