missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Fos Antibody (2H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50037SS
This item is not returnable.
View return policy
Description
c-Fos Monoclonal antibody specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Gene Knock-Out
Specifications
| c-Fos | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Knockout Validated | |
| P01100 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Gene Knock-Out | |
| 2H2 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos | |
| This c-Fos Antibody (2H2) was developed against full length recombinant human c-Fos protein expressed in and purified from E. coli. [UniProt# P01100] | |
| 0.025 mL | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors | |
| 2353 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction