missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Fos Antibody (14C10), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £431.00
Specifications
| Antigen | c-Fos |
|---|---|
| Clone | 14C10 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043907
|
Novus Biologicals
NBP3-26573-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043489
|
Novus Biologicals
NBP3-26573-100ul |
100 μL |
£431.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Fos Monoclonal antibody specifically detects c-Fos in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| c-Fos | |
| Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos | |
| A synthesized peptide derived from Human c-Fos [UniProt P01100] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 14C10 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2353 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title