missing translation for 'onlineSavingsMsg'
Learn More

c-Fms-IN-2, MedChemExpress

Product Code. 16491749
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491749 1 mg 1mg
16463647 50 mg 50mg
16447638 10 mg 10mg
16427546 100 mg 100mg
16420108 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16491749 Supplier Medchem Express Supplier No. HY18787
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress c-Fms-IN-2 is a FMS kinase inhibitor with an IC50 of 0.024 μM.

c-Fms-IN-2 is a FMS kinase inhibitor with an IC50 of 0.024 μM.

Specifications

Chemical Name or Material c-Fms-IN-2
CAS 791587-67-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H21N3O3
Quantity 1 mg
Solubility Information DMSO : ≥ 100 mg/mL (294.65 mM)
SMILES CC1CCN(C2=CC=C(C=C2NC(C3=CC=C(C#N)O3)=O)CO)CC1
Molecular Weight (g/mol) 339.39
Formula Weight 339.39
Percent Purity 95.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.