missing translation for 'onlineSavingsMsg'
Learn More

c-Fms-IN-1, MedChemExpress

Product Code. 16473647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473647 50 mg 50mg
16457638 10 mg 10mg
16437546 100 mg 100mg
16430108 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473647 Supplier Medchem Express Supplier No. HY18791
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress c-Fms-IN-1 is a FMS kinase inhibitor with an IC50 of 0.0008 μM.

c-Fms-IN-1 is a FMS kinase inhibitor with an IC50 of 0.0008 μM.

Specifications

Chemical Name or Material c-Fms-IN-1
Physical Form Solid
CAS 885703-64-0
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H27N5O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 5 mg/mL (12.71 mM; Need ultrasonic)
SMILES CN1CCN(C2=CC=C(NC(C3=CC=C(C#N)O3)=O)C(N4CCCCC4)=C2)CC1
Molecular Weight (g/mol) 393.48
Formula Weight 393.48
Percent Purity 99.4%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.