missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences C-Flex™ Tubing, 374 Formulation

C-Flex™ tubing is manufactured from pharmaceutical-grade thermoplastic elastomer, is fully characterized, validated and tested to a variety of specifications. Extensive biological, chemical, physical properties and extractable testing is available to assist in the validation of C-Flex applications. 

Brand:  Saint Gobain Life Sciences 374-250-2

Product Code. 15660735

  • £117.82 / 15m
Only null left
In Stock
Edge
Add to basket

Description

Description

C-Flex™ tubing is manufactured from pharmaceutical-grade thermoplastic elastomer (TPE) to meet the demands of biopharmaceutical applications for fluid processing with excellent heat seal and sterile weld capabilities.

  • Sealable and weldable either pre- or post- sterilization
  • Sterilizable by gamma irradiation and autoclave
  • Moldable, bondable and formable for single use assemblies and overmolds
  • Complies with USP Class VI and ISO 10993standards
  • Non-pyrogenic, non-cytotoxic, non-hemolytic
  • Remains flexible from -45 to 135°C (-50 to 275°F)
  • Significantly less permeable than silicone
  • Smooth inner bore for superior fluid flow
  • Animal-Derived Component Free (ADCF) and safe for incineration
  • Full Validation Package (biological, chemical, physical, extractables) available for all formulations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP Class VI, ISO 10993, EP
1/4 in.
3/8 in.
50 ft.
0.062 in.
915%
-45°C
Translucent
Flexible
135°C
Autoclavable
6.4 mm
9.5 mm
15.2 m
1.6 mm
Pharmaceutical and biopharmaceutical applications; Ideal for single-use assemblies
Non-sterile
60 (Shore A)
Tensile strength: 1190 psi
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.