Learn More
Ekastu Safety™ Eye Wash Bottle MINI with Funnel
Product Code. 10683043
Change view
Click to view available options
Includes:
Saline Solution
Saline not included
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10683043 Saline Solution Each
10102282 Saline not included Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10683043 Supplier Ekastu Safety™ Supplier No. 177259
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EKASTU Safety™ Eye Wash Bottle MINI with Funnel can be used in both upright and horizontal positions and can be carried in breast and coat bags.
  • Eye Wash Bottle with funnel
  • DIN EN 15154-4
  • Supplied empty or filled
  • For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
  • PE bottle with screwed-on eye funnel and tilting valve
  • Can be used in both upright and horizontal positions
  • leak-proof
  • Stores easily in EPS Eye Wash Station (sold separately)
Specifications
Capacity (Metric) 200 mL
Certifications/Compliance EN 15154-4 DIN
Includes Saline Solution
Material Polyethylene
Product Type Eye Wash Bottle