Learn More
Invitrogen™ C/EBP epsilon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121171
Description
Positive test controls include: NCI-H460, Mouse spleen. The target is usually found in the following locations: Nucleus. Immunogen sequence: MSHGTYYECE PRGGQQPLEF SGGRAGPGEL GDMCEHEASI DLSAYIESGE EQLLSDLFAV KPAPEARGLK GPGTPAFPHY LPPDPRPFAY PPHTFGPDRK ALGPGIYSSP GSYDPRAVAV KEEPRGPEGS RAASRGSYNP LQYQVAHCGQ TAMHLPPTLA APGQPLRVLK APLATAAPPC SPLLKAPSPA.
C/EBP alpha regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. C/EBP alpha uses a bipartite structural motif to bind DNA. Two protein chains dimerize through a set of amphipathic alpha helices termed the leucine zipper. Highly basic polypeptide regions emerge from the zipper to form a linked set of DNA contact surfaces. C/EBP alpha appears to function exclusively in terminally-differentiated, growth-arrested cells. Additional family members include C/EBP beta, C/EBP gamma, C/EBP delta and C/EBP epsilon, all of which exhibit similar DNA-binding specificities and affinities to C/EBP alpha. Furthermore, C/EBP beta and C/EBP delta readily form heterodimers both with each other as well as with C/EBP alpha.
Specifications
| C/EBP epsilon | |
| Polyclonal | |
| Unconjugated | |
| CEBPE | |
| C EBP; c/EBP epsilon; C/EBPe; C/EBPepsilon; C/EBP-epsilon; C/EBP-related protein 1; C/EPBe; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein epsilon; CEBPE; Crp1; Gm294 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1053, 110794 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.88 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15744, Q6PZD9 | |
| CEBPE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human CEBPE (NP_001796.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.