missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C/EBP delta/epsilon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575232
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
C/EBP α regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. C/EBP α uses a bipartite structural motif to bind DNA. Two protein chains dimerize through a set of amphipathic α helices termed the leucine zipper. Highly basic polypeptide regions emerge from the zipper to form a linked set of DNA contact surfaces. C/EBP α appears to function exclusively in terminally-differentiated, growth-arrested cells. Additional family members include C/EBP β, C/EBP γ, C/EBP δ and C/EBP ε, all of which exhibit similar DNA-binding specificities and affinities to C/EBP α. Furthermore, C/EBP β and C/EBP δ readily form heterodimers both with each other as well as with C/EBP α.
Specifications
| C/EBP delta/epsilon | |
| Polyclonal | |
| Unconjugated | |
| CEBPD | |
| C EBP; C/EBP delta; c/EBP epsilon; C/EBPd; C/EBPdelta; C/EBP-delta; C/EBPe; C/EBPepsilon; C/EBP-epsilon; C/EBP-related protein 1; c/EBP-related protein 3; C/EPBe; CCAAT/enhancer binding protein (C/EBP), delta; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein delta; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein delta; CCAAT/enhancer-binding protein epsilon; CEBPD; Cebpe; Celf; CRP1; Crp3; Gm294; NF-IL6-beta; nuclear factor NF-IL6-beta; Transcription factor CELF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1052, 1053, 110794, 12609, 25410, 25695 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P49716, P56261, Q00322, Q03484, Q15744, Q6PZD9 | |
| CEBPD, CEBPE | |
| Synthetic peptide corresponding to amino acids 200-250 of Human C/EBP-delta. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction