Learn More
Invitrogen™ C/EBP beta Recombinant Rabbit Monoclonal Antibody (SC55-05)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532252
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
Specifications
| C/EBP beta | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P17676, P21272, P28033 | |
| Cebpb | |
| Synthetic peptide within Rat C/EBP Beta aa 248-297. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Western Blot, Western Blot | |
| SC55-05 | |
| Unconjugated | |
| Cebpb | |
| AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1051, 12608, 24253 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.