missing translation for 'onlineSavingsMsg'
Learn More

C-DIM12, MedChemExpress

Product Code. p-200097141
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493857

Brand: Medchem Express HY19808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress C-DIM12 is a synthetic Nurr1 activaor induces Nurr1 and DA gene expression in cell lines and primary neurons.

C-DIM12 is a synthetic Nurr1 activaor induces Nurr1 and DA gene expression in cell lines and primary neurons.

Specifications

Chemical Name or Material C-DIM12
Physical Form Solid
CAS 178946-89-9
Color Brown
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H17ClN2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (280.23 mM)
SMILES ClC1=CC=C(C(C2=CNC3=C2C=CC=C3)C4=CNC5=C4C=CC=C5)C=C1
Molecular Weight (g/mol) 356.85
Formula Weight 356.85
Percent Purity 96.61%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.