missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ C-DIM 12
GREENER_CHOICE

Product Code. 30000962
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000962

Brand: Tocris Bioscience™ 5821/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nurr1 activator; inhibits NF-κB-dependent gene expression

Nurr1 activator. Inhibits NF-κB and cytokine expression in microglial cells by stabilizing nuclear corepressor proteins, which reduces binding of p65 to inflammatory gene promoters. Neuroprotective in a mouse Parkinson's disease model. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178946-89-9
Molecular Formula C23H17ClN2

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 356.85
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 356.85
Chemical Name or Material 3,3'-[(4-Chlorophenyl)methylene]bis[1H-indole]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.