missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ c-Di-AMP sodium salt

Click to view available options
Quantity:
702 μg
Unit Size:
702µg
Description
c-Di-AMP sodium salt is an endogenous STING and DDX41 agonist; mediates DDX41-STING interaction. Activates STING-dependent IFN-β production in mouse and human cells. Also A2A inverse agonist. Selectively induces apoptosis of monocytes in human PBMC cultures and in NSG mice transplanted with human CD34+ cells.
This product is sold in units of 702μg, equivalent to 1μmol.
Specifications
Specifications
| Chemical Name or Material | 3′,5′-Cyclic diadenylic acid sodium salt |
| CAS | 2734909-87-4 |
| Quantity | 702 μg |
| Target | STING Antagonists |
| Molecular Formula | C20H22N10Na2O12P2 |
| Formula Weight | 702.38 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction