missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Cbl Recombinant Rabbit Monoclonal Antibody (K01_1P01)

Product Code. 30830421
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830421 50 μL
30825440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830421 Supplier Invitrogen™ Supplier No. MA564763
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

c-Cbl Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Cbl
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1P01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Cbl
Gene Accession No. P22681
Gene Alias 4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL
Gene Symbols Cbl
Host Species Rabbit
Immunogen A synthetic peptide of human CBL.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 867
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.