missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-D Heavy Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £346.00
Specifications
| Antigen | BoNT-D Heavy Chain |
|---|---|
| Clone | 682810 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18741948
![]() |
R&D Systems
MAB6639-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18079441
|
R&D Systems
MAB6639 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BoNT-D Heavy Chain Monoclonal specifically detects BoNT-D Heavy Chain in C. botulinum samples. It is validated for Western Blot.Specifications
| BoNT-D Heavy Chain | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| C. botulinum | |
| P19321 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 682810 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E. coli-derived recombinant Clostridium botulinum Neurotoxin Type D Heavy Chain Asn862-Glu1276 Accession # P19321 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects recombinant C. botulinum BoNT-D Heavy Chain in direct ELISAs and Western blots. 100% and approximately 10% cross-reactivity with recombinant C. botulinum BoNT-C1 Heavy Chain is observed in Western blots and direct ELISAs, respectively. In direct ELISAs, no cross-reactivity with the Heavy Chains of BoNT-A, -E, or -G or the Light Chains of BoNT-A, -B, -C1, -D, -E, -F, -G is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title