missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-A Light Chain Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF4489
This item is not returnable.
View return policy
Description
BoNT-A Light Chain Polyclonal specifically detects BoNT-A Light Chain in C. botulinum samples. It is validated for Western Blot.
Specifications
| BoNT-A Light Chain | |
| Polyclonal | |
| Biotin | |
| Q45894 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5185061 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| atx | |
| E. coli-derived recombinant Clostridium BoNTA Light Chain Met1-Phe425 Accession # Q45894 | |
| 50 μg | |
| Primary | |
| Detects C. botulinum BoNTA Light Chain in Western blots. In Western blots, less than 1% crossreactivity with recombinant C. botulinum BoNTA Heavy Chain is observed. | |
| C. botulinum | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction