missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Abl Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99737CL1
This item is not returnable.
View return policy
Description
c-Abl Polyclonal antibody specifically detects c-Abl in Human samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| c-Abl | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Abelson murine leukemia viral oncogene homolog 1, ABL, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1 | |
| Produced in rabbits immunized with purified, recombinant Human c-Abl (Accession#: NP_009297.2; Pro137-Ser554). | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair, Phospho Specific, Signal Transduction, Tumor Suppressors | |
| 25 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction