Learn More
Ekastu Safety™ C 607/Selecta Model Full Face Mask
Product Code. 10396071
Change view
Click to view available options
Connection Type:
Standard Thread
Unit Size:
Each
Product Code. Connection Type unitSize
10396071 Standard Thread Each
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10396071 Supplier Ekastu Safety™ Supplier No. 466611
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Anti-scratch, anti-fogging, non-reflecting and distortion-free polycarbonate visor with a large field of vision. EKASTU Safety™ C 607/Selecta Model Full Face Mask is constructed of non-irritating EPDM and includes standard thread connection in accordance with DIN EN 148-1.
  • Anti-scratch, anti-fogging, non-reflecting and distortion-free polycarbonate visor with very large field of vision
  • Ultimative level of comfort by ergonomic construction and design
  • Facepiece consists of non-irritating EPDM which is resistant to ageing and guarantees outstanding mechanical and chemical resistance
  • 5-point harness locks the mask quickly and securely in place
  • With standard thread connection DIN EN 148-1
  • 2 Cool Down exhalation valves
Specifications
Certifications/Compliance DIN EN 148-2
Connection Type Standard Thread
Design Type Ergonomic
Material Polycarbonate, EPDM
Product Type Full Face Mask
Harness Style 5-point
Material (Faceshield) Polycarbonate
Material (Lens) Polycarbonate
Quantity 1 Ea.
Weight (Metric) 520 g
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.