Learn More
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing
Product Code. 30053759 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
7/16 in.
Diameter (English) Outer:
1 in.
1-1/4 in.
1/2 in.
1/4 in.
1/8 in.
11/16 in.
13/16 in.
15/16 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
7/16 in.
7/8 in.
9/16 in.
Unit Size:
15m
Product Code. 30053759
Brand: Versilon 5010693
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Tough ether-base polyurethane tubing offers exceptional resistance to oils, grease, and fuels
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Excellent abrasion and tear resistance. Remains flexible even in subzero temperatures. Consistently tight lot-to-lot tolerances. Clear tubing—view fluid being transferred
Specifications
Diameter (English) Inner 3/16 in.
Material Versilon C-210-A
Diameter (English) Outer 5/16 in.
Color Clear