missing translation for 'onlineSavingsMsg'
Learn More

C-176, MedChemExpress

Product Code. 16494728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494728 10 mg 10mg
16457447 5 mg 5mg
16451017 50 mg 50mg
16415556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494728 Supplier Medchem Express Supplier No. HY112906
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress C-176 is a strong and covalent mouse STING inhibitor.

C-176 is a strong and covalent mouse STING inhibitor.

Specifications

Chemical Name or Material C-176
CAS 314054-00-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H7IN2O4
Quantity 10 mg
Solubility Information DMSO : 62.5 mg/mL (174.54 mM; Need ultrasonic)
SMILES O=C(C1=CC=C([N+]([O-])=O)O1)NC2=CC=C(I)C=C2
Molecular Weight (g/mol) 358.09
Formula Weight 358.09
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.