missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-1, Tocris Bioscience™
Protein kinase C inhibitor
Chemical Identifiers
CAS | 84468-24-6 |
---|---|
Molecular Formula | C13H15N3O2S |
Molecular Weight (g/mol) | 277.34 |
MDL Number | MFCD00209852 |
InChI Key | UPTYCYWTFGTCCG-UHFFFAOYSA-N |
Synonym | 1-5-isoquinolinesulfonyl piperazine, ha 100 pharmaceutical, 1-5-isoquinolinylsulfonyl piperazine, chembl75773, 5-piperazine-1-sulfonyl isoquinoline, piperazine, 1-5-isoquinolinylsulfonyl, iqsp, 5-piperazin-1-ylsulfonyl isoquinoline, tocris-0543, c-i-qp |
PubChem CID | 3545 |
IUPAC Name | 5-(piperazine-1-sulfonyl)isoquinoline |
SMILES | O=S(=O)(N1CCNCC1)C1=C2C=CN=CC2=CC=C1 |
Description
Protein kinase inhibitor. Inhibits PKG, PKA and PKC (IC50 values are 4, 8 and 12 μM, respectively). Widely used as a ROCK inhibitor. Increases efficiency of human fibroblast reprogramming with PD 0325901 (Cat.No 4192), CHIR 99021 (Cat.No 4432), A 83-01 (Cat.No 2939) and hLIF. Improves survival of human pluripotent stem cells in culture.Chemical Identifiers
84468-24-6 | |
277.34 | |
UPTYCYWTFGTCCG-UHFFFAOYSA-N | |
3545 | |
O=S(=O)(N1CCNCC1)C1=C2C=CN=CC2=CC=C1 |
C13H15N3O2S | |
MFCD00209852 | |
1-5-isoquinolinesulfonyl piperazine, ha 100 pharmaceutical, 1-5-isoquinolinylsulfonyl piperazine, chembl75773, 5-piperazine-1-sulfonyl isoquinoline, piperazine, 1-5-isoquinolinylsulfonyl, iqsp, 5-piperazin-1-ylsulfonyl isoquinoline, tocris-0543, c-i-qp | |
5-(piperazine-1-sulfonyl)isoquinoline |
Specifications
84468-24-6 | |
MFCD00209852 | |
UPTYCYWTFGTCCG-UHFFFAOYSA-N | |
5-(piperazine-1-sulfonyl)isoquinoline | |
3545 | |
>99% |
C13H15N3O2S | |
1-5-isoquinolinesulfonyl piperazine, ha 100 pharmaceutical, 1-5-isoquinolinylsulfonyl piperazine, chembl75773, 5-piperazine-1-sulfonyl isoquinoline, piperazine, 1-5-isoquinolinylsulfonyl, iqsp, 5-piperazin-1-ylsulfonyl isoquinoline, tocris-0543, c-i-qp | |
O=S(=O)(N1CCNCC1)C1=C2C=CN=CC2=CC=C1 | |
277.34 | |
277.34 | |
C-1 |
Safety and Handling
Recommended Storage : Store at Room Temperature