missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BZW1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92424-0.02ml
This item is not returnable.
View return policy
Description
BZW1 Polyclonal antibody specifically detects BZW1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| BZW1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| basic leucine zipper and W2 domain-containing protein 1, basic leucine zipper and W2 domains 1, basic leucine-zipper protein BZAP45, BZAP45KIAA0005Protein Orf, Nbla10236, putative protein product of Nbla10236 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human BZW1 (NP_001193996.1). MNNQKQQKPTLSGQRFKTRKRDEKERFDPTQFQDCIIQGLTETGTDLEAVAKFLDASGAKLDYRRYAETLFDILVAGGMLAPGGTLADDMMRTDVCVFAAQEDLETMQAFAQVFNKLIRRYKYLEKGFEDEVKKLLLFLKGFSESERNKLAMLTGVLLANGTLNASILNSLYNENLVKEGVSAAFAVKLFKSWINEKDINAVAASLRKVSMDNRLMELFPANKQSVEHFTKYFTEAGLKELSEYVRNQQTIGARKELQKELQEQMSRGDPFKDIILYVKEEMKKNNIPEP | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9689 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction