Learn More
BYSL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28319-1-AP-20UL
This item is not returnable.
View return policy
Description
The BYSL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BYSL. This antibody recognizes human antigen. The BYSL antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BYSL | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 705 | |
| IgG |
| Western Blot, ELISA | |
| 567 μg/mL | |
| Rabbit | |
| Primary | |
| Human |