Learn More
Invitrogen™ BXDC1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570486
Product Code. 15868893
  • £441.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%; Zebrafish: 93%.
RPF2 is a protein coding gene involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BXDC1
Polyclonal
Unconjugated
Rpf2
2810470K21Rik; AU040229; AU043242; bA397G5.4; brix domain containing 1; brix domain-containing protein 1; Brix domain-containing protein 1-like protein; BXDC1; homolog of Rpf2; hypothetical protein LOC406587; ik:tdsubc_1a3; MNCb-4285; ribosomal processing factor 2 homolog; ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; ribosome production factor 2 homolog (S. cerevisiae); RP11-397G5.1; Rpf2; Unknown (protein for MGC:134144); wu:fe17d08; wu:fj16d06; xx:tdsubc_1a3; zgc:77150
Rabbit
Affinity chromatography
RUO
294436
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
0
Rpf2
synthetic peptide directed towards the N-terminal region of rat BXDC1.
100 μL
Primary
Rat
Antibody
IgG
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit