missing translation for 'onlineSavingsMsg'
Learn More

BX795, MedChemExpress

Product Code. 16464018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464018 10 mg 10mg
16442936 25 mg 25mg
16410996 200 mg 200mg
16410377 50 mg 50mg
16406737 5 mg 5mg
16405016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16464018 Supplier Medchem Express Supplier No. HY10514
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BX795 is a potent and selective inhibitor of PDK1, with an IC50 of 6 nM. BX795 is also a potent and relatively specific inhibitor of TBK1 and IKKɛ, with an IC50 of 6 and 41 nM, respectively. BX795 blocks phosphorylation of S6K1, Akt, PKCδ, and GSK3β, and has lower selectivity over PKA, PKC, c-Kit, GSK3β etc. BX795 modulates autophagy.

BX795 is a potent and selective inhibitor of PDK1, with an IC50 of 6 nM. BX795 is also a potent and relatively specific inhibitor of TBK1 and IKKɛ, with an IC50 of 6 and 41 nM, respectively. BX795 blocks phosphorylation of S6K1, Akt, PKCδ, and GSK3β, and has lower selectivity over PKA, PKC, c-Kit, GSK3β etc. BX795 modulates autophagy.

Specifications

Chemical Name or Material BX795
Physical Form Solid
CAS 702675-74-9
Color Light Gray
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H26IN7O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 33.33 mg/mL (56.35 mM; Need ultrasonic)
SMILES O=C(NCCCNC1=C(C=NC(NC2=CC=CC(NC(N3CCCC3)=O)=C2)=N1)I)C4=CC=CS4
Molecular Weight (g/mol) 591.47
Formula Weight 591.47
Percent Purity 98.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.