missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BX471, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BX471 (ZK-811752) is an orally active, potent and selective non-peptide CCR1 antagonist with a Ki of 1 nM, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4.
Specifications
Specifications
| Chemical Name or Material | BX471 |
| CAS | 217645-70-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H24ClFN4O3 |
| Quantity | 10 mg |
| Synonym | ZK-811752 |
| Solubility Information | DMSO : ≥ 100 mg/mL (229.94 mM) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble) |
| SMILES | O=C(N)NC1=CC(Cl)=CC=C1OCC(N2[C@H](C)CN(CC3=CC=C(F)C=C3)CC2)=O |
| Molecular Weight (g/mol) | 434.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction