missing translation for 'onlineSavingsMsg'
Learn More

BVT948, MedChemExpress

Product Code. 16475837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16475837 Supplier Medchem Express Supplier No. HY100625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BVT948 is a protein tyrosine phosphatase (PTP) inhibitor which can also inhibit several cytochrome P450 (P450) isoforms and lysine methyltransferase SETD8 (KMT5A).

BVT948 is a protein tyrosine phosphatase (PTP) inhibitor which can also inhibit several cytochrome P450 (P450) isoforms and lysine methyltransferase SETD8 (KMT5A).

Specifications

Chemical Name or Material BVT948
Physical Form Powder
CAS 39674-97-0
Color Reddish Brown
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C14H11NO3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (414.52 mM; Need ultrasonic)
SMILES O=C1C(O)=C2C(C3=C1C=CC=C3)=NC(C2(C)C)=O
Molecular Weight (g/mol) 241.24
Formula Weight 241.24
Percent Purity 95.3%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.