missing translation for 'onlineSavingsMsg'
Learn More

BVT 2733, MedChemExpress

Product Code. 16463477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463477

Brand: Medchem Express HY18054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BVT 2733 is a potent, selective, and orally active non-steroidal 11β-hydroxydehydrogenase 1 (11β-HSD1) inhibitor. BVT 2733 is potently against the mouse enzyme (IC50=96 nM) over the human enzyme (IC50=3341 nM). BVT 2733 has the potential for the study of arthritis and obesity related disease.

BVT 2733 is a potent, selective, and orally active non-steroidal 11β-hydroxydehydrogenase 1 (11β-HSD1) inhibitor. BVT 2733 is potently against the mouse enzyme (IC50=96 nM) over the human enzyme (IC50=3341 nM). BVT 2733 has the potential for the study of arthritis and obesity related disease.

Specifications

Chemical Name or Material BVT 2733
CAS 376640-41-4
Color Pink
Purity Grade Notes Research
Molecular Formula C17H21ClN4O3S2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (116.56 mM; Need ultrasonic)
SMILES ClC1=C(C)C(S(NC2=NC(CC(N3CCN(C)CC3)=O)=CS2)(=O)=O)=CC=C1
Molecular Weight (g/mol) 428.96
Formula Weight 428.96
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.