missing translation for 'onlineSavingsMsg'
Learn More

BVT-14225, MedChemExpress

Product Code. 16473477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473477 50 mg 50mg
16427458 10 mg 10mg
16427426 100 mg 100mg
16419937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473477 Supplier Medchem Express Supplier No. HY18055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BVT-14225 is a selective 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC50 of 52 nM.

BVT-14225 is a selective 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC50 of 52 nM.

Specifications

Chemical Name or Material BVT-14225
CAS 376638-65-2
Color Pink
Purity Grade Notes Research
Molecular Formula C16H20ClN3O3S2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (248.80 mM)
SMILES O=C(N(CC)CC)CC1=CSC(NS(=O)(C2=CC=CC(Cl)=C2C)=O)=N1
Molecular Weight (g/mol) 401.93
Formula Weight 401.93
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.