missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BVES Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | BVES |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608865
|
Novus Biologicals
NBP2-99427-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659765
|
Novus Biologicals
NBP2-99427-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BVES Polyclonal antibody specifically detects BVES in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| BVES | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| blood vessel epicardial substance, H, HBVES, POP1MGC42413, POPDC1popeye domain containing 1, Popeye domain-containing protein 1, Popeye protein 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human BVES | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 11149 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title