missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BVES Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99427CL1
This item is not returnable.
View return policy
Description
BVES Polyclonal antibody specifically detects BVES in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BVES | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 11149 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| blood vessel epicardial substance, H, HBVES, POP1MGC42413, POPDC1popeye domain containing 1, Popeye domain-containing protein 1, Popeye protein 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human BVES | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction