missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BVDV Envelope glycoprotein E2 Humanized Recombinant Human Monoclonal Antibody (SAA2088)
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554255
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
The BVDV Envelope glycoprotein E2 mediates viral attachment and entry into host cells by binding to cellular receptors.
Specifications
| BVDV Envelope glycoprotein E2 Humanized | |
| Recombinant Monoclonal | |
| 2.58 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| VHH-hFc |
| ELISA | |
| SAA2088 | |
| Unconjugated | |
| Envelope glycoprotein E2 | |
| Recombinant BVDV Envelope glycoprotein E2. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction