missing translation for 'onlineSavingsMsg'
Learn More

BV6, MedChemExpress

Product Code. 16483167
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483167 50 mg 50mg
16449677 5 mg 5mg
16447146 100 mg 100mg
16407148 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483167 Supplier Medchem Express Supplier No. HY16701
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BV6 is an antagonist of cIAP1 and XIAP, members of the inhibitors of apoptosis (IAP) family.

BV6 is an antagonist of cIAP1 and XIAP, members of the inhibitors of apoptosis (IAP) family.

Specifications

Chemical Name or Material BV6
CAS 1001600-56-1
Color White
Purity Grade Notes Research
Molecular Formula C70H96N10O8
Quantity 50 mg
Solubility Information DMSO : ≥ 58 mg/mL (48.11 mM)
SMILES O=C(NCCCCCCNC([C@@H](NC([C@H](CCC1)N1C([C@H](C2CCCCC2)NC([C@H](C)NC)=O)=O)=O)C(C3=CC=CC=C3)C4=CC=CC=C4)=O)[C@@H](NC([C@H](CCC5)N5C([C@H](C6CCCCC6)NC([C@H](C)NC)=O)=O)=O)C(C7=CC=CC=C7)C8=CC=CC=C8
Molecular Weight (g/mol) 1205.57
Formula Weight 1205.57
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.