missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Butyrylcholinesterase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581319
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse Butyrylcholinesterase/BCHE.
Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage.
Specifications
| Butyrylcholinesterase | |
| Polyclonal | |
| Unconjugated | |
| BCHE | |
| acylcholine acylhydrolase; BCHE; BtChE; butylcholinesterase; butyrylcholine esterase; butyrylcholinesterase; C730038G20Rik; CHE1; CHE2; Choline esterase II; cholinesterase; cholinesterase (serum) 2; cholinesterase 1; E1; EC 3.1.1.8; pseudocholinesterase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 12038 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q03311 | |
| BCHE | |
| Recombinant Mouse Butyrylcholinesterase/BCHE Protein (Met1-Leu603). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction