missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyrylcholinesterase/BCHE Antibody (OTI1E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03523
This item is not returnable.
View return policy
Description
Butyrylcholinesterase/BCHE Monoclonal specifically detects Butyrylcholinesterase/BCHE in Human, Canine samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Butyrylcholinesterase/BCHE | |
| Monoclonal | |
| 0.72 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100 | |
| BCHE | |
| Full length human recombinant protein of human BCHE (NP_000046) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 590 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry | |
| OTI1E1 | |
| Unconjugated | |
| Acylcholine acylhydrolase, Butyrylcholine esterase, butyrylcholinesterase, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase | |
| Mouse | |
| 68.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction