missing translation for 'onlineSavingsMsg'
Learn More

Butyrylcarnitine, MedChemExpress

Product Code. 16487487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487487 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16487487 Supplier Medchem Express Supplier No. HY113168
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Butyrylcarnitine is a metabolite in plasma, acts as a biomarker to improve the diagnosis and prognosis of heart failure, and is indicative of anomalous lipid and energy metabolism.

Butyrylcarnitine is a metabolite in plasma, acts as a biomarker to improve the diagnosis and prognosis of heart failure, and is indicative of anomalous lipid and energy metabolism.

Specifications

Chemical Name or Material Butyrylcarnitine
CAS 25576-40-3
Color White
Purity Grade Notes Research
Molecular Formula C11H21NO4
Quantity 5 mg
Solubility Information H2O : ≥ 100 mg/mL (432.36 mM)
SMILES CCCC(O[C@H](CC([O-])=O)C[N+](C)(C)C)=O
Molecular Weight (g/mol) 231.29
Formula Weight 231.29
Percent Purity 97.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.