missing translation for 'onlineSavingsMsg'
Learn More

Butyryl chloride, 99%

Product Code. 10245290 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245290 1 L Glass bottle 1 L
10701744 50 mL Glass bottle 50mL
10153720 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245290 Supplier Thermo Scientific Acros Supplier No. 149210010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-75-3 | C4H7ClO | 106.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-75-3
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000752
InChI Key DVECBJCOGJRVPX-UHFFFAOYSA-N
Synonym butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
PubChem CID 8855
IUPAC Name butanoyl chloride
SMILES CCCC(Cl)=O

Specifications

Melting Point -89.0°C
Color Yellow
Density 1.0180g/mL
Boiling Point 102.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2CH2COCl
Refractive Index 1.4112 to 1.4132
Quantity 1 L
Beilstein 02, 274
Merck Index 15, 1602
Specific Gravity 1.018
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with ether,decomposes in alcohol
Formula Weight 106.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyryl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.